Crotyl chloride, 70%, tech.
Crotyl chloride, 70%, tech.
Crotyl chloride, 70%, tech.
Crotyl chloride, 70%, tech.
Acros Organics
Crotyl chloride, 70%, tech.
CAS: 591-97-9 | C4H7Cl | 90.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
Glass bottle
Catalog number ACR154801000
Price (MYR)
-
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS591-97-9, 563-52-0, 1190-22-3
Specifications
Chemical Name or MaterialCrotyl chloride
Name Notetech., 70%
CAS591-97-9
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.