▸
Molecular Biology Reagents and Kits
▸
DNA Extraction and Purification
▸
Lysing Matrix A, 2 mL Tubes
Actual product may vary
Acros Organics
Lysing Matrix A, 2 mL Tubes
For all sample types except soil for the subsequent isolation of genomic DNA. Lysing Matrix A impact-resistant tubes contain garnet matrix and one 1/4 in. ceramic sphere.
Have Questions?
Catalog Number
Quantity
ACR116910050
50 x 2 mL Tubes
Catalog number
ACR116910050
Price (MYR)
-
Quantity:
50 x 2 mL Tubes
Tube Size:
2mL
Sample Type:
Animal tissues, plants, bacteria, yeast, fungi, algae, insects, nematodes
Molecule to be Enriched:
DNA
Grinding matrix is chemically inert and will not bind nucleic acids
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Specifications
Bead Type
Garnet Matrix, 1/4 in. Ceramic Sphere
Format
Tube
Sample Type
Animal Tissues, Plants, Insects, Worms, Bacteria, Yeast, Fungi, Algae
DNase
Dnase-free
For Use With (Equipment)
FastPrep instruments or other bead beaters
For Use With (Application)
Nucleic Acid Isolation and Purification, Homogenization
Matrix
Lysing Matrix A
Product Type
Lysing matrix
Quantity
50 x 2 mL Tubes
RNase
Rnase-free
Unit Size
Bottle
Contents & Storage
Store at Room Temperature.