Thermo Scientific
N2 Gas Guard
For Use With: 3130/3131, 3140/3141 Incubators
Have Questions?
Catalog NumberFor Use With (Equipment)
1906403130/3131, 3140/3141 incubators
Catalog number 190640
HSN 90328990
Price (MYR)
-
For Use With (Equipment):
3130/3131, 3140/3141 incubators
Minimize the need to frequently change out cylinders and reduce the risk of running out of gas. Thermo Scientific™ Gas Guards for CO2 and LN2 Tanks are factory installed and built in to specified CO2 incubators; specify when ordering the incubator. Fitting unobtrusively inside the incubator casing, these systems interface directly with unit power supply and control systems.
  • Factory-installed systems monitor CO2 or N2 pressure in source tanks
  • Automatically switch from one cylinder to another when supply in first tank is exhausted (pressure falls below 15psi)
  • Audible and visual alarms on incubator control panel alert to tank switch-overs and to when both tanks are low
Specifications
DescriptionGas Guard, CO2
For Use With (Equipment)3130/3131, 3140/3141 incubators
LightingLED
TypeN2 Gas Guard
Unit SizeKit