Grab These Deals Before They're Gone

[Extended Promo] Spotlight Special Offers

Spotlight showcases new special offers on lab products from top brands all year long. Get up to 50% off!

Engage 360°: Campus-Only Discounts

Enjoy exclusive savings of up to 60% on essential lab supplies — available only for our campus community!

[EXTENDED PROMO] Quick Quote

Explore our on-demand quote generation feature called "Quick Quote", which allows you to generate quotes instantly and with ease.

Featured Products

Catalog Number:

A51119700DPC

Thermo Scientific™ LYNX 4000 Superspeed Centrifuge (75008580)

Catalog Number:

75008580

GENESYS™ 140/150 Vis/UV-Vis Spectrophotometers (840-300400)

Catalog Number:

840-300400

Spotlight | Get Up to 50% Off

Enjoy special offers on products from top brands, Save All Year Long! 

Other links

New Lab Setup

Complete planning, equipment, and setup support — all handled by our experts.

Service & Support

Expert care and solutions for your lab operations.

Shop by Brand